Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544055 Supplier Invitrogen™ Supplier No. PA544055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DPVKVIEKKL AIWEQLSPDI KQQYGEGYIE KSLDKLKGNK SYVNMDLSPV Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%.

DHRS9 is a 3-alpha-hydroxysteroid dehydrogenase that converts 3-alpha-tetrahydroprogesterone (allopregnanolone) to dihydroxyprogesterone and 3-alpha-androstanediol to dihydroxyprogesterone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dhrs9
Gene Accession No. Q9BPW9
Gene Alias 3-alpha hydroxysteroid dehydrogenase; 3-alpha-HSD; 3alpha-HSD; C730025I08Rik; dehydrogenase/reductase (SDR family) member 9; dehydrogenase/reductase 9; dehydrogenase/reductase SDR family member 9; DHRS9; epithelial retinol dehydrogenase; eRolDH; NADP-dependent retinol dehydrogenase/reductase; RDH15; RDH-E2; RDHL; RDHTBE; RDH-TBE; Retinol dehydrogenase; retinol dehydrogenase 15; retinol dehydrogenase homolog; RETSDR8; SDR9C4; Short chain dehydrogenase/reductase family 9C member 4; short chain dehydrogenase/reductase family 9C, member 4; short-chain dehydrogenase/reductase retSDR8; Tracheobronchial epithelial cell-specific retinol dehydrogenase; UNQ835/PRO1773
Gene Symbols Dhrs9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DHRS9.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10170
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.