Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHRSX Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297560FR
Description
DHRSX Polyclonal specifically detects DHRSX in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DHRSX | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CXorf11, dehydrogenase/reductase (SDR family) X chromosome, dehydrogenase/reductase (SDR family) X-linked, DHRS5Xmember 1, EC 1.1, SDR46C1 | |
Produced in rabbits immunized with E. coli-derived Human DHRSX fragment. | |
0.1 mL | |
Lipid and Metabolism | |
207063 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction