Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRSX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139939 Supplier Invitrogen™ Supplier No. PA5139939
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LDTLKESGSP GHSARVVTVS SATHYVAELN MDDLQSSACY SPHAAYAQSK LAL Highest antigen sequence indentity to the following orthologs: Mouse - 60%, Rat - 44%.

DHRSX (dehydrogenase/reductase SDR family member on chromosome X) is a 330 amino acid protein belonging to the short-chain dehydrogenases/reductases (SDR) family. Widely expressed, DHRSX is an oxidoreductase that contains a coenzyme binding site and a substrate binding site, indicating a possible role in cellular metabolism. The gene that encodes DHRSX is located in the pseudoautosomal region 1 (PAR1) of X and Y chromosomes. The X and Y chromosomes are the human sex chromosomes. Chromosome X consists of about 153 million base pairs and nearly 1,000 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRSX
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHRSX
Gene Accession No. Q8N5I4
Gene Alias CXorf11; dehydrogenase/reductase (SDR family) X chromosome; dehydrogenase/reductase (SDR family) X-linked; dehydrogenase/reductase (SDR family) Y-linked; dehydrogenase/reductase SDR family member on chromosome X; dehydrogenase/reductase X-linked; DHRS5X; DHRS5Y; DHRSX; DHRSXY; DHRSY; SDR46C1; SDR7C6; Short chain dehydrogenase/reductase family 46C member 1; short chain dehydrogenase/reductase family 46C, member 1; short chain dehydrogenase/reductase family 7C member 6; short chain dehydrogenase/reductase family 7C, member 6; UNQ6508/PRO21433
Gene Symbols DHRSX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHRSX. Recombinant protein control fragment (Product #RP-109517).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207063
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.