Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHTKD1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299197MFV450
Description
DHTKD1 Polyclonal antibody specifically detects DHTKD1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
DHTKD1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
dehydrogenase E1 and transketolase domain containing 1, Dehydrogenase E1 and transketolase domain-containing protein 1, DKFZp762M115, EC 1.2.4.2, KIAA1630DKFZP762M115, MGC3090, probable 2-oxoglutarate dehydrogenase E1 component DHKTD1, mitochondrial | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DHTKD1. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
55526 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction