Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHTKD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598701 Supplier Invitrogen™ Supplier No. PA598701
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHTKD1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The 2-oxoglutarate dehydrogenase complex catalyzes the overall conversion of 2-oxoglutarate to succinyl-CoA and CO(2). It contains multiple copies of three enzymatic components: 2-oxoglutarate dehydrogenase (E1), dihydrolipoamide succinyltransferase (E2) and lipoamide dehydrogenase (E3).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHTKD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DHTKD1
Gene Accession No. A2ATU0, Q96HY7
Gene Alias AMOXAD; C330018I04Rik; CMT2Q; dehydrogenase E1 and transketolase domain containing 1; dehydrogenase E1 and transketolase domain-containing protein 1; Dhtkd1; DKFZp762M115; KIAA1630; MGC3090; Probable 2-oxoglutarate dehydrogenase E1 component DHKTD1, mitochondrial
Gene Symbols DHTKD1
Host Species Rabbit
Immunogen Recombinant Human Probable 2-oxoglutarate dehydrogenase E1 component DHKTD1, mitochondrial protein (1-280AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209692, 55526
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.