Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHX15 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100586AF647
Description
DHX15 Polyclonal antibody specifically detects DHX15 in Human samples. It is validated for Western BlotSpecifications
DHX15 | |
Polyclonal | |
Western Blot | |
ATP-dependent RNA helicase #46, DBP1DEAH box protein 15, DDX15putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15, DEAH (Asp-Glu-Ala-His) box polypeptide 15, EC 3.6.1, EC 3.6.4.13, HRH2DEAD/H box-15, PRP43, PRPF43, PrPp43p, RNA helicase 2 | |
The immunogen recognized by this antibody maps to a region between residue 750 and the C-terminus (residue 795) of human DEAH box polypeptide 15 using the numbering given in Swiss-Prot entry O43143 (GeneID 1665). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
1665 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction