Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541261 Supplier Invitrogen™ Supplier No. PA541261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YINIRKALVT GYFMQVAHLE RTGHYLTVKD NQVVQLHPST VLDHKPEWVL Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

DHX15 (DEAH-Box Helicase 15) is a Protein Coding gene. Among its related pathways are Gene Expression and mRNA Splicing - Major Pathway. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and helicase activity. An important paralog of this gene is DHX8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX15
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DHX15
Gene Accession No. O43143
Gene Alias ATP-dependent RNA helicase #46; DBP1; DDX15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15 (RNA helicase A); DEAD/H box-15; DEAH (Asp-Glu-Ala-His) box helicase 15; DEAH (Asp-Glu-Ala-His) box polypeptide 15; DEAH box protein 15; DEAH9; DEAH-box helicase 15; dhx15; HRH2; im:2639158; mDEAH9; Pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; PRP43; PRPF43; PrPp43p; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15-like protein; RNA helicase 2; wu:fb38f09; wu:fk62f05
Gene Symbols DHX15
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DHX15 (aa 686-735).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1665
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.