Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX15 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501557516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7516 Supplier Bethyl Laboratories, Inc Supplier No. A300390A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Orangutan

DHX15 is a putative ATP-dependent RNA helicase implicated in pre-mRNA splicing.The protein encoded by this gene is a putative ATP-dependent RNA helicase implicated in pre-mRNA splicing.

Specifications

Antigen DHX15
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene DHX15
Gene Accession No. O43143
Gene Alias ATP-dependent RNA helicase No.46, DBP1, DDX15, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15, DEAD/H box-15, DEAH (Asp-Glu-Ala-His) box helicase 15, DEAH (Asp-Glu-Ala-His) box polypeptide 15, DEAH box protein 15, HRH2, PRP43, PRPF43, PrPp43p, putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15, RNA helicase 2
Gene Symbols DHX15
Host Species Rabbit
Immunogen Between 750 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1665
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.