Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX15 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982532
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2532 Supplier Bioss Supplier No. BS13001R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMGB3 (HMG4) belongs to the high mobility group protein superfamily. Like HMG1 and HMG2 , HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription.

Specifications

Antigen DHX15
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DHX15
Gene Accession No. O43143
Gene Alias ATP-dependent RNA helicase #46; BOS_6544; DBP1; DDX15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15 (RNA helicase A); DEAD/H box-15; DEAH (Asp-Glu-Ala-His) box helicase 15; DEAH (Asp-Glu-Ala-His) box polypeptide 15; DEAH box protein 15; Deah9; DEAH-box helicase 15; Dhx15; HRH2; im:2639158; LOC100349473; mDEAH9; Pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; PRP43; PRPF43; PrPp43p; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; RNA helicase 2; wu:fb38f09; wu:fk62f05
Gene Symbols DHX15
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DHX15.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1665
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.