Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHX29 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP185272
Description
DHX29 Polyclonal specifically detects DHX29 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DHX29 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
ATP-dependent RNA helicase DHX29, DDX29, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 29, DEAH (Asp-Glu-Ala-His) box polypeptide 29, DEAH box protein 29, EC 3.6.1, EC 3.6.4.13, FLJ21492, Nucleic acid helicase DDXx | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DHX29 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LQAFSFKTKDIEDAMTNTLLYGGDLHSALDWLCLNLSDDALPEGFSQEFEEQQPKSRPKFQSPQIQATISPPLQPKTK | |
0.1 mL | |
Chromatin Research | |
54505 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction