Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558338 Supplier Invitrogen™ Supplier No. PA558338
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQAFSFKTKD IEDAMTNTLL YGGDLHSALD WLCLNLSDDA LPEGFSQEFE EQQPKSRPKF QSPQIQATIS PPLQPKTK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

DNA helicase and probable main scaffold component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair; according to PubMed:20687897 the contribution to DNA double-strand break repair appears to be largely indirect through transcriptional regulation. Recruited by YY1 to YY1-activated genes, where it acts as an essential coactivator. Binds DNA. In vitro, has double-stranded DNA-dependent ATPase activity. Involved in UV-damage excision repair, DNA replication and chromosome segregation during normal cell division cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX29
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHX29
Gene Accession No. Q7Z478
Gene Alias 3732415M03; ATP-dependent RNA helicase DHX29; AU043276; DDX29; Ddxx; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 29; DEAH (Asp-Glu-Ala-His) box polypeptide 29; DEAH box protein 29; DEAH-box helicase 29; Dhx29; E130202M19Rik; LOC100362324; Nucleic acid helicase DDXx
Gene Symbols DHX29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHX29. Recombinant protein control fragment (Product #RP-96813).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54505
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.