Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541298 Supplier Invitrogen™ Supplier No. PA541298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AESGMAPGGP GEGDGSLVNA SRDLLKEFPQ PKNLLNSVIG RALGISHAKD Sequence homology: Cow: 93%; Dog: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DHX30 is a member of this family.DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The encoded protein has 97% sequence identity with the mouse HELG protein. Alternative splicing of this gene generates 3 transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX30
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DHX30
Gene Accession No. Q7L2E3
Gene Alias 2810477H02Rik; ATP-dependent RNA helicase DHX30; ATP-dependent RNA helicase DHX30; putative ATP-dependent RNA helicase DHX30; C130058C04Rik; Ddx30; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 30; DEAH (Asp-Glu-Ala-His) box helicase 30; DEAH (Asp-Glu-Ala-His) box polypeptide 30; DEAH box protein 30; DEAH-box helicase 30; DExH-box helicase 30; Dhx30; FLJ11214; Helg; KIAA0890; putative ATP-dependent RNA helicase DHX30; RETCOR; Ret-CoR; retina co-repressor; Unknown (protein for MGC:134480)
Gene Symbols DHX30
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DHX30.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22907
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.