Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568736 Supplier Invitrogen™ Supplier No. PA568736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 93%.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The function of this member has not been determined. Alternative splicing of this gene generates 2 transcript variants, but the full length nature of one of the variants has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX32
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DHX32
Gene Accession No. Q7L7V1
Gene Alias 3110079L04Rik; 4732469F02Rik; AA408140; Ddx32; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32; DEAD/H box 32; DEAD/H helicase-like protein 1; DEAD/H helicase-like protein-1; DEAH (Asp-Glu-Ala-His) box polypeptide 32; DEAH box protein 32; DEAH-box helicase 32 (putative); DHLP1; DHX32; helicase DDX32; huDDX32; MuDDX32; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32
Gene Symbols DHX32
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human DHX32.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55760
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.