Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHX33 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002581JF549
Description
DHX33 Polyclonal antibody specifically detects DHX33 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
DHX33 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
DDX33, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 33, DEAH (Asp-Glu-Ala-His) box polypeptide 33, DEAH box protein 33, DKFZp762F2011, EC 3.6.1, EC 3.6.4.13, FLJ21972, putative ATP-dependent RNA helicase DHX33 | |
The immunogen recognized by this antibody maps to a region between residue 650 and the C-terminus (residue 707) of human DEAH (Asp-Glu-Ala-His) Box Polypeptide 33 using the numbering given in entry NP_064547.2 (GeneID 56919). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunofluorescence | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
56919 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction