Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHX33 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567312
Description
Immunogen sequence: ASVMLQLLAMK VPNVLTFDFM SKPSPDHIQA AIAQLDLLGA LEHKDDQLTL TPMGRKMAAF PLEPKFAKTI LMS Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
Stimulates RNA polymerase I transcription of the 47S precursor rRNA. Associates with ribosomal DNA (rDNA) loci where it is involved in POLR1A recruitment. Important element of nucleolar organization.
Specifications
DHX33 | |
Polyclonal | |
Unconjugated | |
DHX33 | |
3110057P17Rik; 9430096J02Rik; ATP-dependent RNA helicase DHX33; DDX33; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 33; DEAH (Asp-Glu-Ala-His) box polypeptide 33; DEAH box protein 33; DEAH-box helicase 33; DHX33; putative ATP-dependent RNA helicase DHX33 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56919 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6R0 | |
DHX33 | |
Recombinant Human DHX33. Recombinant protein control fragment (Product #RP-105584). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction