Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598990
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598990 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598990 Supplier Invitrogen™ Supplier No. PA598990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHX33 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Stimulates RNA polymerase I transcription of the 47S precursor rRNA. Associates with ribosomal DNA (rDNA) loci where it is involved in POLR1A recruitment. Important element of nucleolar organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX33
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DHX33
Gene Accession No. Q9H6R0
Gene Alias 3110057P17Rik; 9430096J02Rik; ATP-dependent RNA helicase DHX33; DDX33; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 33; DEAH (Asp-Glu-Ala-His) box polypeptide 33; DEAH box protein 33; DEAH-box helicase 33; DHX33; putative ATP-dependent RNA helicase DHX33
Gene Symbols DHX33
Host Species Rabbit
Immunogen Recombinant Human ATP-dependent RNA helicase DHX33 protein (116-357AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56919
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.