Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560417 Supplier Invitrogen™ Supplier No. PA560417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVCFRLYAES DYDAFAPYPV PEIRRVALDS LVLQMKSMSV GDPRTFPFIE PPPPASLETA ILYLRDQGAL DSSEALTPIG SLLAQLPVDV V Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX34
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHX34
Gene Accession No. Q14147
Gene Alias 1200013B07Rik; 1810012L18Rik; DDX34; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 34; DEAH (Asp-Glu-Ala-His) box polypeptide 34; DEAH box protein 34; DEAH-box helicase 34; DHX34; HRH1; KIAA0134; mKIAA0134; probable ATP-dependent helicase DHX34; probable ATP-dependent RNA helicase DHX34
Gene Symbols DHX34
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHX34. Recombinant protein control fragment (Product #RP-99084).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9704
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.