Learn More
Invitrogen™ DHX34 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113631
Description
Immunogen sequence: APQDGPPGAE EAALETLQKT SVLQRPYHCE ACGKDFLFTP TEVLRHRKQH For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 87%; Dog: 93%; Guinea Pig: 87%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 87%.
Specifications
| DHX34 | |
| Polyclonal | |
| Unconjugated | |
| DHX34 | |
| 1200013B07Rik; 1810012L18Rik; DDX34; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 34; DEAH (Asp-Glu-Ala-His) box polypeptide 34; DEAH box protein 34; DEAH-box helicase 34; DHX34; HRH1; KIAA0134; mKIAA0134; probable ATP-dependent helicase DHX34; probable ATP-dependent RNA helicase DHX34 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9704 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q14147 | |
| DHX34 | |
| Synthetic peptide directed towards the middle region of human DHX34. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.