Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHX36 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200353F
Description
DHX36 Polyclonal antibody specifically detects DHX36 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DHX36 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DDX36, DEAH (Asp-Glu-Ala-His) box polypeptide 36, DEAH box protein 36, EC 3.6.1, EC 3.6.4.13, G4 resolvase-1, KIAA1488G4R1, MLEL1RHAUDEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36, MLE-like protein 1, probable ATP-dependent RNA helicase DHX36, RNA helicase associated with AU-rich element ARE | |
The immunogen recognized by this antibody maps to a region between residue 950 and the C-terminus (residue 1008) of human DEAH (Asp-Glu-Ala-His) Box Polypeptide 36 using the numbering given in entry NP_065916.1 | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
170506 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction