Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX36 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983248
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3248 Supplier Bioss Supplier No. BS14319R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure. DHX36 (DEAH box protein 36), also known as MLE-like protein 1 and RNA helicase associated with AU-rich element ARE (RHAU), belongs to RNA helicase of the DEAH family and may function in sex development and spermatogenesis. It is expressed in testis and is evolutionary conserved with true orthologs in almost all animal species. DHX36 plays a role in degradation and deadenylation of mRNAs containing in their 3′-UTR the consensus ARE sequence element. DHX36 is required for early embryogenesis.

Specifications

Antigen DHX36
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DHX36
Gene Accession No. Q9H2U1
Gene Alias 2810407E23Rik; AI452301; Asp-Glu-Ala-His; ATP-dependent DNA/RNA helicase DHX36; ATP-dependent RNA helicase DHX36; AU022184; DDX36; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36; DEAD/H box polypeptide 36; DEAH (Asp-Glu-Ala-His) box polypeptide 36; DEAH box protein 36; DEAH-box helicase 36; DEAH-box protein 36; Dhx36; G4 resolvase-1; G4R1; G4-resolvase 1; G4-resolvase-1; Kiaa1488; mKIAA1488; MLEL1; MLE-like protein 1; probable ATP-dependent RNA helicase DHX36; RHAU; RNA helicase associated with AU-rich element ARE; RNA helicase associated with AU-rich element protein
Gene Symbols DHX36
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DHX36.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170506
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.