Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531218
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531218 Supplier Invitrogen™ Supplier No. PA531218
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, HCT116. Predicted reactivity: Mouse (87%), Rat (88%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a DEAD box protein. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX37
Applications Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DHX37
Gene Accession No. Q8IY37
Gene Alias DDX37; DEAD/DEAH box helicase DDX37; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 37; DEAH (Asp-Glu-Ala-His) box polypeptide 37; DEAH box protein 37; DEAH-box helicase 37; DHX37; FLJ41974; Gm1050; Gm451; KIAA1517; LOW QUALITY PROTEIN: probable ATP-dependent RNA helicase DHX37; MGC2695; MGC4322; MGC46245; mKIAA1517; Probable ATP-dependent RNA helicase DHX37
Gene Symbols DHX37
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human DHX37. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57647
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.