Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHX37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541296
Description
Peptide sequence: PLTKKEKKVL QKILEQKEKK SQRAEMLQKL SEVQASEAEM RLFYTTSKLG Sequence homology: Dog: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 79%; Rabbit: 79%; Rat: 93%.
This gene encodes a DEAD box protein. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division.
Specifications
| DHX37 | |
| Polyclonal | |
| Unconjugated | |
| DHX37 | |
| DDX37; DEAD/DEAH box helicase DDX37; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 37; DEAH (Asp-Glu-Ala-His) box polypeptide 37; DEAH box protein 37; DEAH-box helicase 37; DHX37; FLJ41974; Gm1050; Gm451; KIAA1517; LOW QUALITY PROTEIN: probable ATP-dependent RNA helicase DHX37; MGC2695; MGC4322; MGC46245; mKIAA1517; Probable ATP-dependent RNA helicase DHX37 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57647 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IY37 | |
| DHX37 | |
| Synthetic peptide directed towards the N-terminal of human DHX37 (aa 71-120). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction