Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHX38 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040582G
Description
DHX38 Polyclonal antibody specifically detects DHX38 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DHX38 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ATP-dependent RNA helicase DHX38, DDX38pre-mRNA-splicing factor ATP-dependent RNA helicase PRP16, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 38, DEAH (Asp-Glu-Ala-His) box polypeptide 38, DEAH box protein 38, EC 3.6.1, EC 3.6.4.13, KIAA0224pre-mRNA splicing factor ATP-dependent RNA helicase PRP16, PRP16 homolog of S.cerevisiae, PRP16hPrp16, PRPF16 | |
The immunogen recognized by this antibody maps to a region between residue 1150 and 1200 of human DEAH (Asp-Glu-Ala-His) Box Protein 38 using the numbering given in entry NP_054722.2 (GeneID 9785). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9785 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction