Learn More
Invitrogen™ DHX38 Polyclonal Antibody

Description
Antibody detects endogenous levels of total DHX38.
Specifications
Specifications
| Antigen | DHX38 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | DHX38 |
| Gene Accession No. | Q92620 |
| Gene Alias | 5730550P09Rik; AI325984; ATP-dependent RNA helicase DHX38; AW540902; DDX38; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 38; DEAH (Asp-Glu-Ala-His) box polypeptide 38; DEAH box protein 38; DEAH-box helicase 38; DHX38; hPrp16; KIAA0224; mKIAA0224; pre-mRNA splicing factor 16; pre-mRNA splicing factor ATP-dependent RNA helicase PRP16; pre-mRNA-splicing factor ATP-dependent RNA helicase PRP16; PRP16; PRP16 homolog of S.cerevisiae; PRPF16 |
| Gene Symbols | DHX38 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.