Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560685
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560685 Supplier Invitrogen™ Supplier No. PA560685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVGRTFCTMD GRGSPVHIHP SSALHEQETK LEWIIFHEVL VTTKVYARIV CPIRYEWVRD LLPKLHEFNA HDLSSVARRE VREDARRRWT NKENVKQLKD GISKDVLKKM QRRNDDKSIS DARARF Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

This gene encodes a member of the DExH/D box family of ATP-dependent RNA helicases that have an essential role in RNA metabolism. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX40
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHX40
Gene Accession No. Q8IX18
Gene Alias 2410016C14Rik; ARG147; DDX40; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 40 (RNA helicase); DEAH (Asp-Glu-Ala-His) box polypeptide 40; DEAH box protein 40; DEAH-box helicase 40; Dhx40; PAD; probable ATP-dependent RNA helicase DHX40; Protein PAD
Gene Symbols DHX40
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHX40. Recombinant protein control fragment (Product #RP-93191).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79665
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.