Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546142 Supplier Invitrogen™ Supplier No. PA546142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KQLKDGISKD VLKKMQRRND DKSISDARAR FLERKQQRTQ DHSDTRKETG Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Rat: 86%.

This gene encodes a member of the DExH/D box family of ATP-dependent RNA helicases that have an essential role in RNA metabolism. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX40
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DHX40
Gene Accession No. Q8IX18
Gene Alias 2410016C14Rik; ARG147; DDX40; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 40 (RNA helicase); DEAH (Asp-Glu-Ala-His) box polypeptide 40; DEAH box protein 40; DEAH-box helicase 40; Dhx40; PAD; probable ATP-dependent RNA helicase DHX40; Protein PAD
Gene Symbols DHX40
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DHX40 (aa 730-779).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79665
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.