Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHX57 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128693DL594
Description
DHX57 Polyclonal antibody specifically detects DHX57 in Human samples. It is validated for Western BlotSpecifications
DHX57 | |
Polyclonal | |
Western Blot | |
DDX57, DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57, DEAH box protein 57, DEAH-box RNA/DNA helicase AAM73547, EC 3.6.1, EC 3.6.4.13, FLJ32861, putative ATP-dependent RNA helicase DHX57 | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57 using the numbering given in entry NP_945314.1 (GeneID 90957). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
90957 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction