Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHX57 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568742
Description
This target displays homology in the following species: Cow: 87%; Dog: 93%; Guinea Pig: 100%; Horse: 79%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Probable ATP-binding RNA helicase.
Specifications
DHX57 | |
Polyclonal | |
Unconjugated | |
DHX57 | |
AW494914; DDX57; DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57; DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57 homolog; DEAH box protein 57; DEAH-box helicase 57; DEAH-box RNA/DNA helicase AAM73547; DExH-box helicase 57; DHX57; Putative ATP-dependent RNA helicase DHX57 | |
Rabbit | |
Affinity chromatography | |
RUO | |
90957 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6P158 | |
DHX57 | |
synthetic peptide directed towards the C terminal region of human DHX57. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction