Learn More
DHX57 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302299A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Probable ATP-binding RNA helicase.Specifications
DHX57 | |
Polyclonal | |
Unconjugated | |
DHX57 | |
DDX57, DEAH box protein 57, DEAH-box RNA/DNA helicase AAM73547, putative ATP-dependent RNA helicase DHX57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90957 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6P158 | |
DHX57 | |
Between 50 and 100 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.