Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561923 Supplier Invitrogen™ Supplier No. PA561923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDGSLSQAAM MQSALAKERR ELKQAQREAE MDSIPMGLNK HWVDPLPDAE GRQIAANMRG IGMMPNDIPE WKKHAFGGNK ASYGKKTQMS ILEQRESLP Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene is a member of the DEAH box polypeptide family. The encoded protein contains the DEAH (Asp-Glu-Ala-His) motif which is characteristic of all DEAH box proteins, and is thought to function as an ATP-dependent RNA helicase that regulates the release of spliced mRNAs from spliceosomes prior to their export from the nucleus. This protein may be required for the replication of human immunodeficiency virus type 1 (HIV-1). Alternative splicing results in multiple transcript variants. [provided by RefSeq, Oct 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dhx8
Gene Accession No. Q14562
Gene Alias ATP-dependent RNA helicase DHX8; DDX8; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 8; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 8 (RNA helicase); DEAH (Asp-Glu-Ala-His) box polypeptide 8; DEAH box protein 8; DEAH-box helicase 8; DHX8; HRH1; LOW QUALITY PROTEIN: ATP-dependent RNA helicase DHX8; LRRGT00035; mDEAH6; mKIAA4096; PRP22; PRPF22; RNA helicase; RNA helicase HRH1
Gene Symbols Dhx8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHX8. Recombinant protein control fragment (Product #RP-101103).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1659
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.