Learn More
DHX9 Monoclonal Antibody (8E3), Active Motif™

Description
Specifications
Specifications
| Antigen | DHX9 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 8E3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | Dhx9 |
| Gene Accession No. | O70133, Q08211 |
| Gene Alias | AI326842; ATP-dependent RNA helicase A; BOS_16077; Ddx9; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9; DEAH (Asp-Glu-Ala-His) box helicase 9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DEAH box protein 9; DEAH-box helicase 9; DExH-box helicase 9; Dhx9; FLJ17406; HEL-5; Leukophysin; LKP; mHEL-5; NDH II; NDH2; NDHII; Nuclear DNA helicase II; OTTHUMP00000033287; RHA; RNA helicase; RNA helicase A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.