Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHX9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115120 Supplier Invitrogen™ Supplier No. PA5115120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DHX9.

The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E.coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Dhx9
Gene Accession No. O70133, Q08211
Gene Alias AI326842; ATP-dependent RNA helicase A; Ddx9; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9; DEAH (Asp-Glu-Ala-His) box helicase 9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DEAH box protein 9; DEAH-box helicase 9; DExH-box helicase 9; Dhx9; FLJ17406; HEL-5; Leukophysin; LKP; mHEL-5; NDH II; NDH2; NDHII; Nuclear DNA helicase II; OTTHUMP00000033287; RHA; RNA helicase; RNA helicase A
Gene Symbols Dhx9
Host Species Rabbit
Immunogen A synthesized peptide derived from human DHX9(Accession Q08211), corresponding to amino acid residues S1139-Y1189.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13211, 1660, 304859
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.