Learn More
DHX9 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300854A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Bovine and Orangutan
This gene encodes a member of the DEAH-containing family of RNA helicases. The encoded protein is an enzyme that catalyzes the ATP-dependent unwinding of double-stranded RNA and DNA-RNA complexes. This protein localizes to both the nucleus and the cytoplasm and functions as a transcriptional regulator. This protein may also be involved in the expression and nuclear export of retroviral RNAs. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 11 and 13.Specifications
DHX9 | |
Polyclonal | |
Unconjugated | |
DHX9 | |
DHX9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DDX9; LKP; NDHII; RHA; ATP-dependent RNA helicase A; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A); DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A, nuclear DNA helicase II; leukophysin); DEAD/H box-9 (nuclear DNA helicase II; RNA helicase A); OTTHUMP00000033287; leukophysin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1660 | |
4° C | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q08211 | |
DHX9 | |
Between 200 and 250 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.