Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di(2-thienyl)-1,1,2,2-tetramethyldisilane, 97%, Thermo Scientific™

Catalog No. AAH2893803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2893803 1 g
AAH2893806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2893803 Supplier Thermo Scientific Chemicals Supplier No. H2893803

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124733-24-0
Molecular Formula C12H18S2Si2
Molecular Weight (g/mol) 282.57
MDL Number MFCD09266191
InChI Key FGYSQQKHYAVZEZ-UHFFFAOYSA-N
Synonym di 2-thienyl-1,1,2,2-tetramethyldisilane, 1,1,2,2-tetramethyl-1,2-bis thiophen-2-yl disilane, bis 2-thienyl-1,1,2,2-tetramethyldisilane, amtsi088, 1,1,2,2-tetramethyl-1,2-di 2-thienyl disilane, 1,1,2,2-tetramethyl-1,2-di thiophen-2-yl disilane
PubChem CID 14382824
IUPAC Name [dimethyl(thiophen-2-yl)silyl]-dimethyl-thiophen-2-ylsilane
SMILES C[Si](C)(C1=CC=CS1)[Si](C)(C)C1=CC=CS1

Specifications

Melting Point 42°C to 44°C
Quantity 1 g
Formula Weight 282.57
Percent Purity 97%
Chemical Name or Material Di(2-thienyl)-1,1,2,2-tetramethyldisilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.