Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-µ-chlorobis(p-cymene)chlororuthenium(II), 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
52462-29-0
C20H28Cl4Ru2
612.39
MFCD00064793
LAXRNWSASWOFOT-UHFFFAOYSA-J
dichloro p-cymene ruthenium ii dimer,di-mu-chloro-bis chloro p-cymene ruthenium ii,rucl2 p-cymene 2,ru p-cymene cl2 2,dichloro p-cymene ruthenium dimer,p-cymene ruthenium ii chloride dimer,dichloro-p-cymene ruthenium ii dimer
10908223
dichlororuthenium;1-methyl-4-propan-2-ylbenzene
CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl
52462-29-0
C20H28Cl4Ru2
612.39
MFCD00064793
LAXRNWSASWOFOT-UHFFFAOYSA-J
dichloro p-cymene ruthenium ii dimer,di-mu-chloro-bis chloro p-cymene ruthenium ii,rucl2 p-cymene 2,ru p-cymene cl2 2,dichloro p-cymene ruthenium dimer,p-cymene ruthenium ii chloride dimer,dichloro-p-cymene ruthenium ii dimer
10908223
dichlororuthenium;1-methyl-4-propan-2-ylbenzene
CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl
Melting Point 250°C
Elemental Analysis 97.5% min.
Packaging Glass bottle
Quantity 1g
Color Brown-Red to Orange-Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble
Formula Weight 612.39
Physical Form Crystals or Powder
Percent Purity 98%
Chemical Name or Material Di-μ-chlorobis(p-cymene)chlororuthenium(II)
Show More Show Less

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc

GHS Signal Word: Danger