Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ DI Liners for Shimadzu GCs
SDP

Catalog No. 06718611 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5/Pk.
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-718-611 5/Pk.
06-718-597 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-718-611 Supplier Restek™ Supplier No. 21719
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Shimadzu 17A, 2010, and 2014 GCs equipped with split/splitless inlets.

  • Trace, dirty, high MW, active samples, high recovery and linearity
  • Geometry: Uniliner
  • Material: Borosilicate Glass
  • Deactivation: Intermediate Polarity (IP)
  • Packing: Deactivated Wool
  • I.D.: 3.5mm
  • OD: 5mm
  • Length: 95mm

Specifications

Product Type IP Uniliner Inlet Liner
Packing Material Deactivated Wool
Length (Metric) 9.5 cm
Quantity 5/Pk.
Diameter (Metric) Inner 0.35 cm
Diameter (Metric) Outer 0.5 cm
For Use With (Equipment) Shimadzu 17 A/2010/2014 GCs w/ Split/Splitless Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.