Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Di-Methyl Lysine Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116818
Description
Positive Samples: NIH/3T3, C6, Mouse liver, Rat liver, HeLa, MCF7.
Methylated lysine is derivative of the amino acid residue lysine where the sidechain ammonium group has been methylated one or more times.
Specifications
Di-Methyl Lysine | |
Polyclonal | |
Unconjugated | |
Dimethyllysine | |
A synthetic peptide corresponding to a sequence containing dimethylated K. | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, Western Blot | |
0.50 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Chemical, Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction