Learn More
Di-^m-bromobis(tri-tert-butylphosphine)dipalladium(I)
Description
Coupling reactions. Will activate aryl chloride and sterically hindered or electron rich aryl/vinyl bromides and iodides. Especially active in difficult aminations.Di-μ-bromobis(tri-tert-butylphosphine)dipalladium(I) is used as a catalyst for Suzuki coupling, Negishi coupling and Buchwald-hartwig amination reactions. It is used as a catalyst for C-C, C-N and C-S bond formation, gamma-arylation of alfa,beta-unsaturated esters and diastereoselective arylation of 4-substituted cyclohexyl esters. It is also involved in aromatic halide substitution reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Coupling reactions
- Will activate aryl chloride and sterically hindered or electron rich aryl/vinyl bromides and iodides
- Especially active in difficult aminations

Specifications
Specifications
Color | Green |
Linear Formula | [Pd(C4H9)3PBr]2 |
Quantity | 0.5 g |
Sensitivity | Air sensitive |
Solubility Information | Soluble in benzene and tolueneSoluble in tetrahydrofuran,toluene,dichloromethane and chloroform; Insoluble in alcohols and water. |
Formula Weight | 777.29 |
Physical Form | Powder |
Chemical Name or Material | Di-mu-bromobis (tri-tert-butylphosphine)dipalladium (I) |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Store at -20°C; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.