Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-^m-chlorodichlorobis(ethylene)diplatinum(II), 97%

Catalog No. AA3928503
Click to view available options
Quantity:
250 mg
1 g

CAS: 12073-36-8 | C4H8Cl4Pt2 | 588.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12073-36-8
Molecular Formula C4H8Cl4Pt2
Molecular Weight (g/mol) 588.08
MDL Number MFCD00058713
InChI Key XBLBDYCZFZESPN-UHFFFAOYSA-J
Synonym di-mu-chloro-dichlorobis ethylene diplatinum ii, zeise's dimer, ptcl2 c2h4 2, di-chloro-dichlorobis ethylene diplatinum ii, di-micro-chlorodichlorobis ethylene diplatinum ii
PubChem CID 11124780
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Pt++].[Pt++].C=C.C=C

Specifications

Quantity 1 g
IUPAC Name diplatinum(2+) bis(ethene) tetrachloride
Formula Weight 588.1
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Di-mu-chlorodichlorobis(ethylene)diplatinum(II)
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 235-135-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.