Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-butyl phthalate, 99+%

Catalog No. AA3124414
Click to view available options
Quantity:
5 g
25 g
1 kg
3 kg

CAS: 84-74-2 | C16H22O4 | 278.35 g/mol

Dibutyl phthalate (DBP) is a common industrial chemicals used as plasticizers, softeners, adhesives or solvents used by a variety of industries. It is also used as an additive to adhesives or printing inks. Other applications include in PVC, paints, cosmetics and coatings of cars.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-74-2
Molecular Formula C16H22O4
Molecular Weight (g/mol) 278.35
MDL Number MFCD00009441
InChI Key DOIRQSBPFJWKBE-UHFFFAOYSA-N
Synonym dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db
PubChem CID 3026
ChEBI CHEBI:34687
IUPAC Name 1,2-dibutyl benzene-1,2-dicarboxylate
SMILES CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC

Specifications

Melting Point -35°C
Density 1.045 g/mL
Boiling Point 339°C to 340°C
Flash Point 171°C (340°F)
Odor Odorless
Refractive Index 1.492
Quantity 25 g
UN Number UN3082
Beilstein 1914064
Merck Index 14,3035
Solubility Information Very soluble in alcohol, ether, acetone, benzene
Formula Weight 278.35
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Di-n-butyl phthalate
Show More Show Less
Hazard Category H336-H360Df-H373
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 201-557-4
RTECSNumber TI0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.