Alfa Aesar™ Di-n-butyl phthalate, 99+%

Manufacturer: Alfa Aesar™ 3124406

Quantity 5g

 View more versions of this product

Catalog No. AA3124406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 84-74-2
    Formula Weight 278.35
    MDL Number MFCD00009441
    UN Number UN3082
    Beilstein 1914064
    Merck Index 14,3035
    Chemical Name or Material Di-n-butyl phthalate

    Specifications

    Formula Weight 278.35
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 1914064
    RTECS Number TI0875000
    Density 1.045g/mL
    Melting Point −35°C
    Boiling Point 339° to 340°C
    Odor Odorless
    Refractive Index 1.4920
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Quantity 5g
    Physical Form Liquid
    MDL Number MFCD00009441
    CAS 84-74-2
    UN Number UN3082
    Merck Index 14,3035
    Solubility Information Very soluble in alcohol, ether, acetone, benzene
    Health Hazard 1 Danger
    TSCA TSCA
    Flash Point 171° (340°F)
    Health Hazard 2 GHS H Statement
    H360
    May damage fertility or the unborn child.
    Chemical Name or Material Di-n-butyl phthalate
    Assay ≥99%

    GHS H Statement
    H360
    May damage fertility or the unborn child.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit