Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Di-n-butylamine, for HPLC
Click to view available options
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -62.0°C |
| Color | White |
| Density | 0.7600g/mL |
| Boiling Point | 159.0°C |
| Flash Point | 39°C |
| Packaging | Glass bottle |
| Refractive Index | 1.4160 to 1.4180 |
| Linear Formula | [CH3(CH2)3]2NH |
| Quantity | 10 mL |
| Residue after Evaporation | 0.0005% max. |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. Toxic if swallowed. Toxic in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fre |
| EINECSNumber | 203-921-8 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction