Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-butyltin bis(2-ethylhexanoate)

Catalog No. AA7112614
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 2781-10-4 | C24H48O4Sn | 519.35 g/mol

Di-n-butyltin bis(2-ethylhexanoate) is used in silanol condensation reactions for caulk and sealant applications and in the production of silicones. It is also used in one or two component systems which are compatible with aromatic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2781-10-4
MDL Number MFCD00015261

Specifications

Melting Point 57°C to 59°C
Density 1.070 g/mL
Boiling Point 220°C (2 mmHg)
Odor Odorless
Molecular Formula C24H48O4Sn
Quantity 25 g
UN Number UN3146
Synonym dibutyltin bis 2-ethylhexanoate, dibutyltin di 2-ethylhexoate, stannane, dibutylbis 2-ethyl-1-oxohexyl oxy, di-n-butyltin di-2-ethylhexanoate, di-n-butyltin bis 2-ethylhexanoate, dibutyltin di-2-ethylhexanoate, dibutyltin di 2-ethylhexanoate, dibutylbis 2-ethyl-1-oxohexyl oxy stannane, tin, dibutyl-, di 2-ethylhexoate, dibutyltin di-2-ethylhexoate
Solubility Information Soluble in mineral oil, acetone, xylene
InChI Key GPDWNEFHGANACG-UHFFFAOYNA-L
SMILES CCCCC(CC)C(=O)O[Sn](CCCC)(CCCC)OC(=O)C(CC)CCCC
IUPAC Name dibutyl[(2-ethylhexanoyl)oxy]stannyl 2-ethylhexanoate
Molecular Weight (g/mol) 519.35
Formula Weight 519.34
Physical Form Lumps
Chemical Name or Material Di-n-butyltin bis (2-ethylhexanoate)
Show More Show Less
Hazard Category H301-H340-H360-H372-H500
Hazard Statement GHS H Statement
H301-H310-H361
Toxic if swallowed.
Fatal in contact with skin.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 220-481-2
RTECSNumber WH6714500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.