Learn More
Di-n-butyltin dichloride, 96%
Description
Organotin intermediate.Di-n-butyltin dichloride is used as an organotin intermediate. It serves as heat and light stabilizer for polyvinyl chloride plastics. It is also used in the preparation of methyl 2-(methoxycarbonylmethyl)-5-iodobenzene by reacting with 2,5-diiodobenzoic acid. It acts as an esterification catalyst and a veterinary vermicide and tapeworm remedy. Further, it is used as ion exchange agent, process regulators and processing aids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 39°C to 41°C |
| Color | White |
| Density | 1.36 g/mL |
| Boiling Point | 135°C (10 mmHg) |
| Flash Point | 168°C (334°F) |
| Odor | Acrid |
| Refractive Index | 1.4991 |
| Linear Formula | [CH3(CH2)3]2SnCl2 |
| Quantity | 250 g |
| UN Number | UN2928 |
| Show More |
Safety and Handling
| Hazard Category | H301-H312-H314-H330-H341-H360FD-H372 |
| Hazard Statement | GHS H Statement H301-H330-H360-H372-H341-H314-H312 Toxic if swallowed. Fatal if inhaled. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Causes severe skin burns and eye damage. Harmful in contact with skin. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S. |
| EINECSNumber | 211-670-0 |
| RTECSNumber | WH7100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.