Learn More
Di-n-butyltin dilaurate, 95%
Description
Di-n-butyltin dilaurate is used as a catalyst for the production of polyurethanes as well as for the transesterification reactions. It is involved in the vulcanization of silicones and a stabilizer in polyvinyl chloride (PVC). It acts as a rust inhibitor for polyurethanes, polyols, silicones and as a fuel additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | <10°C |
Color | Yellow |
Density | 1.066 g/mL |
Boiling Point | >204°C (12 mmHg) |
Flash Point | 235°C (455°F) |
Refractive Index | 1.473 |
Linear Formula | [CH3(CH2)3]2Sn[O2C(CH2)10CH3]2 |
Quantity | 100 g |
UN Number | UN2788 |
Merck Index | 14,3038 |
Show More |
Safety and Handling
Hazard Category | H314-H317-H335-H336-H341-H360FD-H370-H372 |
Hazard Statement | GHS H Statement H301-H331-H360-H341-H314-H318-H317 Toxic if swallowed. Toxic if inhaled. May damage fertility or the unborn child. Suspected of causing genetic defects. Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 201-039-8 |
RTECSNumber | WH7000000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.