Learn More
(Di-n-dodecyl)dimethylammonium bromide, 98+%
Description
(Di-n-dodecyl)dimethylammonium bromide is used as a phase transfer catalyst. It is also used as floor disinfectant. Further, it acts as surface-active agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(Di-n-dodecyl)dimethylammonium bromide is used as a phase transfer catalyst. It is also used as floor disinfectant. Further, it acts as surface-active agent.
Solubility
Soluble in hot and cold water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 157°C to 162°C |
| Linear Formula | [CH3(CH2)11]2N(CH3)2Br |
| Quantity | 2 g |
| Beilstein | 3576662 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in hot and cold water. |
| Formula Weight | 462.65 |
| Percent Purity | ≥98% |
| Chemical Name or Material | (Di-n-dodecyl)dimethylammonium bromide |
Safety and Handling
| Hazard Category | H302-H315-H319-H335-H500 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 221-923-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.