Learn More
Di-n-octyl phthalate, 98%
Description
Di-n-octyl phthalate is used as a plasticizer for many resins and elastomers. It acts as an additive. It is also used for medical tubing and blood storage bags, wire and cables, carpet back coating, floor tile and in cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.98g/mL |
| Assay Percent Range | 98% |
| Quantity | 25 mL |
| Merck Index | 14,2864 |
| Formula Weight | 390.56 |
| Percent Purity | 98% |
| Physical Form | Liquid |
| Chemical Name or Material | Di-n-octyl phthalate |
Safety and Handling
| RTECSNumber | TI1925000 |
| TSCA | TSCA |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.