Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-octyldichlorotin, 95%

Catalog No. aa4173806
Click to view available options
Quantity:
5 g
25 g

CAS: 3542-36-7 | C16H36Cl2Sn | 418.07 g/mol

Di-n-octyldichlorotin compounds are used as intermediates in the production of other organotin chemicals. Metatin and Acima. These products are also used as catalysts or industrial intermediates in the production of polyurethanes, silicone resins, synthetic lubricants, and MS polymer sealants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3542-36-7
Molecular Formula C16H36Cl2Sn
Molecular Weight (g/mol) 418.07
MDL Number MFCD00018091
InChI Key DXPJPPSKHFECLR-UHFFFAOYSA-N
Synonym dichlorodioctyltin, dioctyltin dichloride, di-n-octyltin dichloride, stannane, dichlorodioctyl, dotc, di-n-octyltindichloride, dichloro dioctyl stannane, di-n-octyldichlorotin, stannane, dioctyldichloro, di-n-octyl-zinn dichlorid
PubChem CID 77080
IUPAC Name dichloro(dioctyl)stannane
SMILES Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC

Specifications

Melting Point 44°C to 48°C
Boiling Point 175°C (1 mmHg)
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 416.04
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Di-n-octyldichlorotin
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 222-583-2
RTECSNumber WH7247000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.