Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine hydrochloride, 99.5%, without anticaking agent

Catalog No. AC364790100
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle

CAS: 50-01-1 | CH5N3·HCl | 95.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH5N3·HCl
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Absorbance (6M aq. soln.), (6 M aq. soln.) at 0nm, 0.01 max. at 280nm, 0.03 max. at 260nm
Melting Point 178.0°C to 188.0°C
Color Colorless to White
Density 1.3450g/mL
Infrared Spectrum conform to structure and standard
Assay Percent Range 99.5%
Packaging Plastic Drum
Linear Formula H2NC(=NH)NH2·HCl
Quantity 10 kg
Beilstein 03,82
Merck Index 15,4597
Solubility Information Solubility in water: 2150g/L (20°C).
Formula Weight 95.53
Percent Purity 99.5%
Physical Form Crystals
Chemical Name or Material Guanidine Hydrochloride, Without anticaking agent
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse
EINECSNumber 200-002-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.