Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Di-n-propyl sulfide, 98+%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.
Solubility
Soluble in water 351mg/L @ 25°C.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.
Solubility
Soluble in water 351mg/L @ 25°C.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | -102°C |
Density | 0.838 |
Boiling Point | 141°C to 142°C |
Flash Point | 28°C (82°F) |
Odor | Stench |
Refractive Index | 1.4487 |
Quantity | 100 g |
UN Number | UN1993 |
Beilstein | 1719002 |
Merck Index | 14,7868 |
Show More |
Safety and Handling
Hazard Category | H226-H315-H319-H335 |
Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
EINECSNumber | 203-873-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction